N-Boc-4-methyl-L-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-p-Me-Phe-OH, Boc-Phe(4-Me)-OH

OR14572-5G
APOSOR14572-5GEA 79.9 EUR
APOSOR14572-5G
N-Boc-4-methyl-L-phenylalanine
N-Boc-4-methyl-L-phenylalanine
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.336 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317027
CAS-Nummer: 80102-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR