Boc-4-methyl-D-phenylalanine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-D-Phe(4-Me)-OH, Boc-p-Me-D-Phe-OH
OR14573-5G
APOSOR14573-5GEA
126
EUR
APOSOR14573-5G
Boc-4-methyl-D-phenylalanine 98%
Boc-4-methyl-D-phenylalanine
Formel:
C₁₅H₂₁NO₄ Molecular Weight: 279.336 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01317726 CAS-Nummer: 80102-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...