Boc-D-Tic-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

OR14588-250MG
APOSOR14588-250MGEA 21.9 EUR
APOSOR14588-250MG
Boc-D-Tic-OH
Boc-D-Tic-OH
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143818
CAS-Nummer: 115962-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR