Fmoc-D-Tic-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

OR14590-25G
APOSOR14590-25GEA 81.2 EUR
APOSOR14590-25G
Fmoc-D-Tic-OH
Fmoc-D-Tic-OH
Formel: C₂₅H₂₁NO₄
Molecular Weight: 399.446 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00144345
CAS-Nummer: 130309-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR