Fmoc-2,4-dichloro-D-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: 2,4-Dichloro-D-phenylalanine, N-FMOC protected

OR14605-5G
APOSOR14605-5GEA 183 EUR
APOSOR14605-5G
Fmoc-2,4-dichloro-D-phenylalanine 97%
Fmoc-2,4-dichloro-D-phenylalanine
Formel: C₂₄H₁₉Cl₂NO₄
Molecular Weight: 456.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863047
CAS-Nummer: 352351-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR