Fmoc-3,4-dichloro-D-phenylalanine 95+%
Lieferant: Apollo Scientific
Synonyme:
Fmoc-D-Phe(3,4-DiCl)-OH, Fmoc-3,4-dichloro-D-Phe-OH
OR14609-1G
APOSOR14609-1GEA
33.5
EUR
APOSOR14609-1G
Fmoc-3,4-dichloro-D-phenylalanine 95+%
Fmoc-3,4-dichloro-D-phenylalanine
Formel:
C₂₄H₁₉Cl₂NO₄ Molecular Weight: 456,319 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273472 CAS-Nummer: 177966-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...