N-Fmoc-4-iodo-D-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-D-Phe(4-I)-OH

OR14613-5G
APOSOR14613-5GEA 82.8 EUR
APOSOR14613-5G
N-Fmoc-4-iodo-D-phenylalanine
N-Fmoc-4-iodo-D-phenylalanine
Formel: C₂₄H₂₀INO₄
Molecular Weight: 513.331 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00672557
CAS-Nummer: 205526-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR