(2-Chlor-3-pyridinyl)methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR1462-100G
APOSOR1462-100GEA
140
EUR
APOSOR1462-100G
(2-Chlor-3-pyridinyl)methanol 97%
(2-Chlor-3-pyridinyl)methanol
Formel:
C₆H₆ClNO Schmelzpunkt: 54…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01935961 CAS-Nummer: 42330-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...