Fmoc-L-3,3-diphenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-diphenylpropanoic acid, Fmoc-β,β-diphenyl-Ala-OH

OR14628-1G
APOSOR14628-1GEA 87 EUR
APOSOR14628-1G
Fmoc-L-3,3-diphenylalanine 97%
Fmoc-L-3,3-diphenylalanine
Formel: C₃₀H₂₅NO₄
Molecular Weight: 463,533 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672337
CAS-Nummer: 201484-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR