Fmoc-L-4,4'-biphenylalanine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-4-phenyl-Phe-OH, Fmoc-Bip(4,4')-OH

OR14632-10G OR14632-1G OR14632-5G
APOSOR14632-10GEA 215 EUR
APOSOR14632-10G APOSOR14632-1G APOSOR14632-5G
Fmoc-L-4,4'-biphenylalanine 95%
Fmoc-L-4,4'-biphenylalanine
Formel: C₃₀H₂₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00191198
CAS-Nummer: 199110-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR