Fmoc-L-4-pyridylalanine 98%

Lieferant: Apollo Scientific

Synonyme: N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH

OR14664-5G
APOSOR14664-5GEA 120 EUR
APOSOR14664-5G
Fmoc-L-4-pyridylalanine 98%
Fmoc-L-4-pyridylalanine
Formel: C₂₃H₂₀N₂O₄
Molecular Weight: 388.43 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00672566
CAS-Nummer: 169555-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR