(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

OR14679-5G
APOSOR14679-5GEA 119 EUR
APOSOR14679-5G
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95+%
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229.276 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01320858
CAS-Nummer: 161660-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR