N-(4-Brombutyl)phthalimid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Brombutyl)-1,2-benzoldicarbonsäureimid, N-(4-Brombutyl)phthalsäureimid
OR1468-100G
APOSOR1468-100GEA
88.6
EUR
APOSOR1468-100G
N-(4-Brombutyl)phthalimid 98%
N-(4-Brombutyl)phthalimid
Formel:
C₁₂H₁₂BrNO₂ Schmelzpunkt: 76…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005905 CAS-Nummer: 5394-18-3 EINECS: 226-401-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...