Fmoc-7-Ahp-OH 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-7-aminoheptanoic acid

OR14683-5G
APOSOR14683-5GEA 91.5 EUR
APOSOR14683-5G
Fmoc-7-Ahp-OH 98%
Fmoc-7-Ahp-OH
Formel: C₂₂H₂₅NO₄
Molecular Weight: 367,445 g/mol
Schmelzpunkt: 152…157 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01861353
CAS-Nummer: 127582-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR