2-Brom-L-phenylalanin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Phe(2-Br)-OH
OR14693-1G
APOSOR14693-1GEA
21.9
EUR
APOSOR14693-1G
2-Brom-L-phenylalanin 97%
2-Brom-L-phenylalanin
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.1 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03092958 CAS-Nummer: 42538-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...