4-Brom-L-phenylalanin 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
H-Phe(4-Br)-OH
OR14695-10G
APOSOR14695-10GEA
91.5
EUR
APOSOR14695-10G
4-Brom-L-phenylalanin 95+%
4-Brom-L-phenylalanin
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.1 g/mol Schmelzpunkt: 265 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063062 CAS-Nummer: 24250-84-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...