D-Homophenylalanin 98%

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-4-phenylbutyric Acid, H-D-HomoPhe-OH, H-D-HoPhe-OH

OR14698-25G
APOSOR14698-25GEA 84.1 EUR
APOSOR14698-25G
D-Homophenylalanin 98%
D-Homophenylalanine
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063091
CAS-Nummer: 82795-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR