3-Nitro-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(3-nitrophenyl)propanoic acid, H-Phe(3-NO₂)-OH

OR14707-1G OR14707-5G
APOSOR14707-1GEA 75.4 EUR
APOSOR14707-1G APOSOR14707-5G
3-Nitro-L-phenylalanine 97%
3-Nitro-L-phenylalanine
Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01860661
CAS-Nummer: 19883-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR