3-(1-Naphthyl)-D-alanine

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-1-Nal-OH

OR14712-1G
APOSOR14712-1GEA 21.9 EUR
APOSOR14712-1G
3-(1-Naphthyl)-D-alanine
3-(1-Naphthyl)-D-alanine
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215,252 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00038544
CAS-Nummer: 78306-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR