3-(2-Pyridyl)-D-alanine

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-2-Pal-OH, H-β-(2-Pyridyl)-D-Ala-OH

OR14722-250MG
APOSOR14722-250MGEA 40.6 EUR
APOSOR14722-250MG
3-(2-Pyridyl)-D-alanine
3-(2-Pyridyl)-D-alanine
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237245
CAS-Nummer: 37535-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR