(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-3-D-Pal-OH, 3-(Pyridin-3-yl)-D-alanine, (2R)-2-Amino-3-(pyridin-3-yl)propionic acid, 3-Aza-D-phenylalanine

OR14724-250MG
APOSOR14724-250MGEA 26.1 EUR
APOSOR14724-250MG
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00079673
CAS-Nummer: 70702-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR