3-(2-Thienyl)-L-alanine 95%

Lieferant: Apollo Scientific

Synonyme: H-β-(2-Thienyl)-Ala-OH, 3-(2-Thienyl)-L-alanine, (S)-2-Amino-3-(thiophen-2-yl)propanoic acid

OR14732-5G
APOSOR14732-5GEA 150 EUR
APOSOR14732-5G
3-(2-Thienyl)-L-alanine 95%
3-(2-Thienyl)-L-alanine
Formel: C₇H₉NO₂S
Molecular Weight: 171.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064335
CAS-Nummer: 22951-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR