2-Allyl-D-glycine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
D-Allylglycine, H-D-Allyl-Gly-OH, H-D-Gly(Allyl)-OH
OR14737-5G
APOSOR14737-5GEA
109
EUR
APOSOR14737-5G
2-Allyl-D-glycine 98%
2-Allyl-D-glycine
Formel:
C₅H₉NO₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063104 CAS-Nummer: 54594-06-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...