3-Fur-2-yl-D-alanine 95+%

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-3-(furan-2-yl)propanoic acid, H-D-2-Furyl-Ala-OH

OR14739-250MG
APOSOR14739-250MGEA 29 EUR
APOSOR14739-250MG
3-Fur-2-yl-D-alanine 95+%
3-Fur-2-yl-D-alanine
Formel: C₇H₉NO₃
Molecular Weight: 155,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860879
CAS-Nummer: 110772-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR