3-(4,6-Dimethoxypyrimidin-2-yl)propanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR1474-1G OR1474-5G
APOSOR1474-1GEA 255 EUR
APOSOR1474-1G APOSOR1474-5G
3-(4,6-Dimethoxypyrimidin-2-yl)propanoic acid 95+%
3-(4,6-Dimethoxypyrimidin-2-yl)propanoic acid
Formel: C₉H₁₂N₂O₄
Molecular Weight: 212.21 g/mol
Schmelzpunkt: 120…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01568075
CAS-Nummer: 386715-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR