1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR14748-10G OR14748-1G OR14748-5G
APOSOR14748-10GEA 0 EUR
APOSOR14748-10G APOSOR14748-1G APOSOR14748-5G
1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride
1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride
Formel: C₈H₁₉Cl₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00023304
CAS-Nummer: 2031-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR