1,4-Dihydroxy-2,6-dimethoxybenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dimethoxybenzene-1,4-diol

OR14749-1G
APOSOR14749-1GEA 39.3 EUR
APOSOR14749-1G
1,4-Dihydroxy-2,6-dimethoxybenzene
1,4-Dihydroxy-2,6-dimethoxybenzene
Formel: C₈H₁₀O₄
Molecular Weight: 170,17 g/mol
Schmelzpunkt: 158…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017160
CAS-Nummer: 15233-65-5
EINECS: 239-282-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR