2,4-Dichloro-6-methylpyrimidin-5-ylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichloro-6-methylpyrimidin-5-amine, 5-Amino-2,4-dichloro-6-methylpyrimidine

OR14760-1G
APOSOR14760-1GEA 21.9 EUR
APOSOR14760-1G
2,4-Dichloro-6-methylpyrimidin-5-ylamine
2,4-Dichloro-6-methylpyrimidin-5-ylamine
Formel: C₅H₅Cl₂N₃
Molecular Weight: 178.02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08062586
CAS-Nummer: 13162-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR