(3,5-Dimethylisoxazol-4-yl)acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(3,5-Dimethylisoxazol-4-yl)acetic acid

OR14801-1G
APOSOR14801-1GEA 95.7 EUR
APOSOR14801-1G
(3,5-Dimethylisoxazol-4-yl)acetic acid 98%
(3,5-Dimethylisoxazol-4-yl)acetic acid
Formel: C₇H₉NO₃
Molecular Weight: 155,15 g/mol
Schmelzpunkt: 120…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05864546
CAS-Nummer: 2510-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR