DL-O-Methylserin

Lieferant: Apollo Scientific
Achtung

OR14804-1G
APOSOR14804-1GEA 0 EUR
APOSOR14804-1G
DL-O-Methylserin
DL-O-Methylserine
Formel: C₄H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00037773
CAS-Nummer: 19794-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR