2-(4-Aminophenyl)propanenitrile

Lieferant: Apollo Scientific
Achtung

OR14806-250MG
APOSOR14806-250MGEA 65.4 EUR
APOSOR14806-250MG
2-(4-Aminophenyl)propanenitrile
2-(4-Aminophenyl)propanenitrile
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Schmelzpunkt: 55…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09865026
CAS-Nummer: 28694-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR