2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR14814-1G
APOSOR14814-1GEA
270
EUR
APOSOR14814-1G
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
2-[(4-Chlorobenzyl)amino]benzenecarboxylic acid
Formel:
C₁₄H₁₂ClNO₂ Molecular Weight: 261,71 g/mol Schmelzpunkt: 146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03032901 CAS-Nummer: 14345-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...