2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid

Lieferant: Apollo Scientific

OR14825-1G
APOSOR14825-1GEA 270 EUR
APOSOR14825-1G
2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid
2,4-Dioxo-1,4-dihydro-2H-3,1-benzoxazine-8-carboxylic acid
Formel: C₉H₅NO₅
Molecular Weight: 207,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09865033
CAS-Nummer: 167902-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR