3-Ethoxy-2-pyridinamine

Lieferant: Apollo Scientific
Achtung

OR14829-1G
APOSOR14829-1GEA 336 EUR
APOSOR14829-1G
3-Ethoxy-2-pyridinamine
3-Ethoxy-2-pyridinamine
Formel: C₇H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00234961
CAS-Nummer: 10006-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR