5-Amino-3-isopropyl-1,2,4-thiadiazol

Lieferant: Apollo Scientific
Achtung

OR14831-250MG
APOSOR14831-250MGEA 65.4 EUR
APOSOR14831-250MG
5-Amino-3-isopropyl-1,2,4-thiadiazol
5-Amino-3-isopropyl-1,2,4-thiadiazol
Formel: C₅H₉N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD03093824
CAS-Nummer: 32039-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR