Methyl-1,3-benzodioxole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl benzo[d][1,3]dioxole-5-carboxylate

OR14837-1G
APOSOR14837-1GEA 33.5 EUR
APOSOR14837-1G
Methyl-1,3-benzodioxole-5-carboxylate
Methyl-1,3-benzodioxole-5-carboxylate
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030273
CAS-Nummer: 326-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR