4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Acetylpiperazine-1-carboxamidine hydroiodide

OR1484-1G
APOSOR1484-1GEA 183 EUR
APOSOR1484-1G
4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide
4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide
Formel: C₇H₁₅IN₄O
Molecular Weight: 298.13 g/mol
Schmelzpunkt: 229…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797494
CAS-Nummer: 849776-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR