(6-Methyl-3-pyridinyl)methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline

OR14843-25G
APOSOR14843-25GEA 220 EUR
APOSOR14843-25G
(6-Methyl-3-pyridinyl)methanol
(6-Methyl-3-pyridinyl)methanol
Formel: C₇H₉NO
Molecular Weight: 123.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08692003
CAS-Nummer: 34107-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR