5-Amino-3-phenyl-1,2,4-thiadiazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Phenyl-1,2,4-thiadiazol-5-amine

OR14852-250MG
APOSOR14852-250MGEA 21.9 EUR
APOSOR14852-250MG
5-Amino-3-phenyl-1,2,4-thiadiazol 95%
5-Amino-3-phenyl-1,2,4-thiadiazole
Formel: C₈H₇N₃S
Molecular Weight: 177,23 g/mol
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003159
CAS-Nummer: 17467-15-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR