5-Methylthien-2-yl isocyanate 97%

Lieferant: Apollo Scientific
Gefahr

OR14873-1G OR14873-250MG OR14873-500MG
APOSOR14873-1GEA 203 EUR
APOSOR14873-1G APOSOR14873-250MG APOSOR14873-500MG
5-Methylthien-2-yl isocyanate 97%
5-Methylthien-2-yl isocyanate

May form small amount of sediment

Formel: C₆H₅NOS
Siedepunkt: 94 °C (53 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD09817547
CAS-Nummer: 76536-99-7
UN: 2206
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR