5-Bromo-1-methylindoline 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-bromo-1-methylindoline, 5-Bromo-2,3-dihydro-1-methyl-1H-indole

OR14883-1G
APOSOR14883-1GEA 0 EUR
APOSOR14883-1G
5-Bromo-1-methylindoline 90%
5-Bromo-1-methylindoline
Formel: C₉H₁₀BrN
Molecular Weight: 212,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09817498
CAS-Nummer: 99848-78-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR