15-Hydroxypentadecansäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR1489-1G
APOSOR1489-1GEA
31.9
EUR
APOSOR1489-1G
15-Hydroxypentadecansäure 99%
15-Hydroxypentadecanoic acid
Formel:
C₁₅H₃₀O₃ Molecular Weight: 258.40 g/mol Schmelzpunkt: 84…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039535 CAS-Nummer: 4617-33-8 EINECS: 225-026-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...