[2-(Thien-2-yl)pyrimidin-5-yl]methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[5-(Hydroxymethyl)pyrimidin-2-yl]thiophene, 5-(Hydroxymethyl)-2-(thien-2-yl)pyrimidine
OR14895-250MG
APOSOR14895-250MGEA
232
EUR
APOSOR14895-250MG
[2-(Thien-2-yl)pyrimidin-5-yl]methanol 97%
[2-(Thien-2-yl)pyrimidin-5-yl]methanol
Formel:
C₉H₈N₂OS Molecular Weight: 192,238 g/mol Schmelzpunkt: 115,5…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09863233 CAS-Nummer: 921939-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...