7-(Aminomethyl)-1-methyl-1H-indole 97%

Lieferant: Apollo Scientific
Gefahr

OR14910-100MG
APOSOR14910-100MGEA 0 EUR
APOSOR14910-100MG
7-(Aminomethyl)-1-methyl-1H-indole 97%
7-(Aminomethyl)-1-methyl-1H-indole
Formel: C₁₀H₁₂N₂
Lagertemperatur: Kühlschrank
MDL: MFCD09879961
CAS-Nummer: 937795-97-6
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR