4-(3-Bromothien-2-yl)phenacyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethan-1-one

OR14920-250MG
APOSOR14920-250MGEA 174 EUR
APOSOR14920-250MG
4-(3-Bromothien-2-yl)phenacyl bromide 97%
4-(3-Bromothien-2-yl)phenacyl bromide
Formel: C₁₂H₈Br₂OS
Molecular Weight: 360,064 g/mol
Schmelzpunkt: 45…47,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879969
CAS-Nummer: 944450-89-9
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR