4-((2-(Bromomethyl)phenyl)sulphonyl)morpholine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[(Morpholin-4-yl)sulphonyl]benzyl bromide

OR14926-100MG
APOSOR14926-100MGEA 36.4 EUR
APOSOR14926-100MG
4-((2-(Bromomethyl)phenyl)sulphonyl)morpholine 95%
4-((2-(Bromomethyl)phenyl)sulphonyl)morpholine
Formel: C₁₁H₁₄BrNO₃S
Molecular Weight: 320,2 g/mol
Schmelzpunkt: 71,5…73.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879984
CAS-Nummer: 941717-06-2
UN: 3261
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR