7-Bromo-1-methyl-1H-indole 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR14929-250MG
APOSOR14929-250MGEA
21.9
EUR
APOSOR14929-250MG
7-Bromo-1-methyl-1H-indole 95%
7-Bromo-1-methyl-1H-indole
Formel:
C₉H₈BrN Schmelzpunkt: 50,5…53 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09879960 CAS-Nummer: 280752-68-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...