2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR14933-1G
APOSOR14933-1GEA
87
EUR
APOSOR14933-1G
2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
2-Morpholino-1,3-thiazole-5-carbaldehyde
Formel:
C₈H₁₀N₂O₂S Schmelzpunkt: 162,5…164.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01568825 CAS-Nummer: 1011-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...