2-(Thien-2-yl)pyrimidine-5-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR14935-250MG
APOSOR14935-250MGEA 92.8 EUR
APOSOR14935-250MG
2-(Thien-2-yl)pyrimidine-5-carboxaldehyde 97%
2-(Thien-2-yl)pyrimidine-5-carboxaldehyde
Formel: C₉H₆N₂OS
Molecular Weight: 190,222 g/mol
Schmelzpunkt: 183…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817553
CAS-Nummer: 921939-12-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR