4-(Hydroxymethyl)-4-(thien-2-yl)tetrahydro-2H-pyran

Lieferant: Apollo Scientific

Synonyme: (4-Thien-2-yltetrahydropyran-4-yl)methanol

OR14941-1G
APOSOR14941-1GEA 296 EUR
APOSOR14941-1G
4-(Hydroxymethyl)-4-(thien-2-yl)tetrahydro-2H-pyran
4-(Hydroxymethyl)-4-(thien-2-yl)tetrahydro-2H-pyran
Formel: C₁₀H₁₄O₂S
Molecular Weight: 198,282 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879932
CAS-Nummer: 906352-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR